Learn More
Heidolph™ Hei-SHAKE Micro Reaction Vessel Attachment with 20 Positions
SDP
Catalog No. 12100028 Shop All Heidolph North America Products
Change view
Click to view available options
Autoclavable:
Autoclavable
Catalog No. Autoclavable
12-100-028 Autoclavable
1 options
Catalog No. 12-100-028 Supplier Heidolph™ Supplier No. 036213735
Add to Cart
Add to Cart
Micro reaction vessel attachment for Hei-SHAKE Orbital Core platform shaker.
Attachment or rack for holding 20 micro reaction vessels such as Eppis 1.5 -2.0 mL.
  • Material: stainless steel (frame) / polyoxymethylene (inner part)
  • Characteristics: autoclavable up to 121°C and dishwasher safe; angle adjustment up to 90° attachment without foot part can be used as table frame
This attachment is an optional accessory, must be ordered separately with the base 12-100-026.
Specifications
Autoclavable Autoclavable
Color Silver
Holds Micro Reaction Vessels
Material Stainless Steel (Frame), POM (Inner)
Positions 20
Shape Rectangular
Stackable No
Sterility Non-sterile
Product Type Vessel Rack
Shelves 1
Array 10 x 2