Learn More
Heidolph™ Hei-VAC Control
SDP
Catalog No. 02321071 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei-VAP Evaporators
Catalog No. For Use With (Equipment)
02-321-071 Hei-VAP Evaporators
1 options
Catalog No. 02-321-071 Supplier Heidolph™ Supplier No. 036308001
Add to Cart
Add to Cart
Heidolph™ Hei-VAC Control for accurate vacuum control of house vacuum systems or valve regulated vacuum pumps
  • For accurate vacuum control of house vacuum systems or valve regulated vacuum pumps
  • Affordable option for digital vacuum control
  • Easy fixation on the condenser shaft above the evaporator or as stand alone device with base next to the unit
  • Manual Vent button prevents bumping
  • Graphs in order display the entire vacuum process
  • RS232 interface
  • Multilingual menu navigation
  • Easy installation and operation
  • Compatible with all Hei-VAP Evaporators which do not include an integrated vacuum control
  • Vacuum controller complete with integrated ceramic vacuum sensor, regulation valve and venting valve
  • With wall-plug power supply
  • Ready for use, with manual
Specifications
Connection Type RS232 Interface
Description Multilingual Navigation Menu
Type Vacuum Controller
For Use With (Equipment) Hei-VAP Evaporators
Includes Wall-Plug Power Supply
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov