Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Receiving Flasks
SDP
Catalog No. 13878707 Shop All Heidolph North America Products
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Type:
Coated Receiving Flask
Standard Receiving Flask
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Type
13-878-707 250 mL Standard Receiving Flask
13-878-714 2000 mL Standard Receiving Flask
13-878-575 1000 mL Coated Receiving Flask
13-878-706 100 mL Coated Receiving Flask
13-878-716 3000 mL Standard Receiving Flask
13-878-710 500 mL Coated Receiving Flask
13-878-715 2000 mL Coated Receiving Flask
13-878-705 100 mL Standard Receiving Flask
13-878-574 1000 mL Standard Receiving Flask
13-878-717 3000 mL Coated Receiving Flask
13-878-708 250 mL Coated Receiving Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 13-878-707 Supplier Heidolph™ Supplier No. 036302500
Add to Cart
Add to Cart
Coated or standard receiving flasks
Standard and Surlyn-coated receiving flasks for Heidolph Hei-VAP Rotary Evaporator
  • NS 35/20 joint size
Specifications
Type Standard Receiving Flask
For Use With (Equipment) Hei-VAP Rotary Evaporator
Taper 35/20
Volume (Metric) 250 mL
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov