Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Tubing
SDP
Catalog No. 02321079 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Tubing
Tubing Set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
02-321-079 Tubing
13-878-675 Tubing Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-321-079 Supplier Heidolph™ Supplier No. 036294800
Add to Cart
Add to Cart
Accessory parts and components for Heidolph Hei-VAP Rotary Evaporators.
  • Tubing is for vacuum and water connection
  • Made of Polyethylene 940 Material
Specifications
Material Polyethylene
Type Tubing
For Use With (Equipment) Hei-Vap™ Rotary Evaporator
Includes Individual 1 m Piece
Length (Metric) 1 m
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov