Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Hei-VOLUME DISTIMATIC PRO
SDP

Catalog No. 13687434 Shop All Heidolph North America Products
Change view
Click to view available options
Voltage:
100/240 V
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Voltage
13-687-434 100/240 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-687-434 Supplier Heidolph™ Supplier No. 036064500

To request a quote for this item, please select Request a Quote to reach our dedicated support team.

missing translation for 'validMainframeMsg'

Automatic module for continuous distillation and optimal solvent management.

Hei-VOLUME Distimatic Pro: Automatic Module for Continuous Distillation

The Hei-VOLUME Distimatic Pro automatic module enables efficient, unattended operation for high-volume rotary evaporation across a wide range of applications. In daily workflows involving solvent concentration, recovery, processing, or recycling, tasks like filling, draining, and cleaning often consume the most time. Automation offers a way to boost throughput without increasing labor or system footprint. The Distimatic Pro uses advanced sensors and pressure-controlled fluid transfer to fully automate filling and draining, supporting both large-batch and continuous process workflows.

The Distimatic can be paired with either benchtop Hei-VAP rotary evaporators or process-sale Hei-VAP Industrial Large Scale rotary evaporators.

Automated Functions:

  • Filling and refilling of the rotary flask
  • Distillate drainage via overpressure
  • Automatic shutdown of all system components at process end or in case of disturbance
  • Integrated cleaning program for glass components and media-carrying hoses

Application Examples:

  • Pharmaceutical and Contract Labs
    • Solvent recovery after chromatography
    • Solvent recycling for cleaning processes
  • Analysis and Research Labs
    • Concentration of water samples
    • Sample preparation for study of rainwater radioactivity
  • Food Industry
    • Solvent recovery from stevia extracts
    • Concentration in lactose crystallization
  • Beverage Production
    • Botanical extraction for gin production
  • Plant Extraction
    • Extraction of medical plants for analysis
    • Solvent recovery after plant extraction
  • Nuclear Power Plants
    • Wastewater treatment
    • Concentration of water samples
  • Automotive Manufacturing
    • Recycling of cleaning fluids

Specifications

Certifications/Compliance CE, TUV, NRTL
Temperature Range 5°C to 31°C (80% RH), 32°C to 40°C (50% RH)
Type Immersion Circulator
Display Detachable digital control panel, 7 in. touch panel, two control knobs
For Use With (Application) Hei-VAP Industrial Rotary Evaporator, A-series glassware
Includes Distimatic Pro, Distimatic Pro Support, Vacuum unit consisting of mounting plate 1 and mounting plate 2, glassware consisting of buffer vessel, collector vessel, educt sensor vessel, distributor, condenser coupling including fastening clamps, tubing set
Model Automatic Hei-VOLUME Distimatic Pro
Timer 1 to 9999 sec.
Warranty 3 Years
Dimensions (D x W x H) 521 x 186 x 429 mm
Width (English) 7.32 in.
Width (Metric) 186 mm
Height (English) 16.89 in.
Height (Metric) 429 mm
Depth (English) 20.51 in.
Depth (Metric) 521 mm
Weight (English) 37.48 lb.
Weight (Metric) 17 kg
Electrical Requirements 100/240 V, 50/60 Hz
Voltage 100/240 V
Frequency 50/60 Hz
Connection Type L+N+PE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.