Learn More
Heidolph™ Hei-TORQUE Overhead Stirrer Flexible Shaft
SDP
Catalog No. 13880154 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Flexible Shaft
Catalog No. Type
13-880-154 Flexible Shaft
1 options
Catalog No. 13-880-154 Supplier Heidolph™ Supplier No. 036300600
Add to Cart
Add to Cart
Flexible Shaft for use with Hei-TORQUE Overhead Stirrer.
  • For use with all RZR Overhead Stirrers except RZR 2102 control Z
  • Chuck included
Specifications
For Use With (Equipment) Hei-TORQUE Core, Hei-TORQUE Expert, Hei-TORQUE Ultimate
Includes Chuck
Type Flexible Shaft
Weight (Metric) 80 oz.
Weight (English) 5 lb.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov