Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ G5 Glassware Replacement Part - O-ring Seal
SDP

Catalog No. 13878626 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
G5 Glassware
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-878-626 G5 Glassware
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-878-626 Supplier Heidolph™ Supplier No. 036301690
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Accessory for G5 Dry Ice Glassware Set

  • Orange o-ring
  • For low boiling liquids

Specifications

For Use With (Application) Low Boiling Liquids
For Use With (Equipment) G5 Glassware
Product Type O-Ring
Color Orange
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.