Learn More
Heidolph™ Glassware - G5B Glass Set Complete
GSA_VASDP
Catalog No. 13878639 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Laborota Interchangeable Glassware
Catalog No. For Use With (Equipment)
13-878-639 Laborota Interchangeable Glassware
1 options
Catalog No. 13-878-639 Supplier Heidolph™ Supplier No. 036301820
Add to Cart
Add to Cart
Request a Quote
For low-boiling solvents
  • Glassware set includes on 1000mL evaporating flask and one 1000mL receiving flask
  • Transparent plastic coating (Halar-coated) for added safety
  • Evaporating flasks and vapor tubes which come with a standard NS 24/40 joint size are also available with NS 24/32 ground joints
  • All glassware sets feature GL 10 thread; the inlet tube can be connected with a threaded fitting
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type G5B Dry Ice Condenser
For Use With (Equipment) Laborota Interchangeable Glassware
Material (Coating) Transparent Plastic
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov