Learn More
Heidolph™ Hei-Vap™ Glassware - G5 Glass Set
SDP
Catalog No. 13878637 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Dry Ice Condenser for Low-Boiling Solvents
Catalog No. For Use With (Equipment)
13-878-637 Dry Ice Condenser for Low-Boiling Solvents
1 options
Catalog No. 13-878-637 Supplier Heidolph™ Supplier No. 036301800
Add to Cart
Add to Cart
Request a Quote
Dry-Ice condenser for low-boiling solvents
  • Glassware set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All glassware sets feature GL 10 thread; the inlet tube can be connected with a threaded fitting
  • All glassware sets are also available with transparent plastic halar coating for G5B and abcite coating for G1B/G3B/G6B
  • Glassware sets G3 and G6 allow for the automated distillation program AUTOaccurate
Specifications
Type Glassware Set G5
For Use With (Equipment) Dry Ice Condenser for Low-Boiling Solvents
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov