Learn More
Heidolph™ Hei-Vap™ Rotary Evaporator Accessories - Foam Brakes
SDP
Catalog No. 13878703 Shop All Heidolph North America Products
Change view
Click to view available options
Taper:
24/40
29/32
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Taper
13-878-703 29/32
13-878-704 24/40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-878-703 Supplier Heidolph™ Supplier No. 036302460
Add to Cart
Add to Cart
For use with Heidolph Hei-VAP series rotary evaporators
  • The foam brake prevents contents of the evaporating flask from being drawn into the condenser in the event of bumping.
Specifications
Type Foam Brake
For Use With (Equipment) Hei-VAP Rotary Evaporator
Includes Flask clamp
Taper 29/32
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov