Learn More
Heidolph™ Hei-Vap Rotary Evaporator Temperature Sensors
SDP
Catalog No. 13889340 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
AUTOaccurate Sensor
Vapor Temperature Sensor
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-889-340 Vapor Temperature Sensor
13-889-341 AUTOaccurate Sensor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-889-340 Supplier Heidolph™ Supplier No. 036308010
Add to Cart
Add to Cart
Accessory parts and components for Heidolph Hei-VAP Rotary Evaporators
AUTOaccurate-sensor
  • For Hei-VAP Precision, Hei-VAP Expert Control, and Hei-VAP Ultimate Control
  • Only in combination with glassware set G3 and G6
Vapor Temperature Sensor
  • For Hei-VAP Advantage Hei-VAP Precision, Hei-VAP Expert, Hei-VAP Ultimate, Hei-VAP Expert Control, and Hei-VAP Ultimate Control
Specifications
Type Vapor Temperature Sensor
For Use With (Equipment) Hei-VAP rotary evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov