Learn More
Heidolph™ Immersion Blenders - SilentCrusher M Accessories: Vessel Holding Bracket
SDP
Catalog No. 13900808 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
SilentCrusher M Generator
Catalog No. For Use With (Equipment)
13-900-808 SilentCrusher M Generator
1 options
Catalog No. 13-900-808 Supplier Heidolph™ Supplier No. 036300860
Accessories for use with Heidolph SilentCrusher M blender
For holding vessels used with SilentCrusher M Generator.
  • For vessel diameters 20 to 105mm
  • Includes 2 retainer rings
Specifications
Product Type Holding Bracket
For Use With (Application) Ideal for holding vessels
For Use With (Equipment) SilentCrusher M Generator
Includes 2 retainer rings