Learn More
Heidolph™ Rotary Evaporator Accessories - Interchangeable Glassware
SDP
Catalog No. 13878570 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
1000 mL Evaporating Flask
Clamp S 35
G1 Diagonal Condenser with safety coating
G3 Vertical Condenser
Jacket for Condensate Trap, G5 Condenser
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
13-878-570 G1 Diagonal Condenser with safety coating
13-878-578 Clamp S 35
13-878-585 G3 Vertical Condenser
13-878-623 Jacket for Condensate Trap, G5 Condenser
13-878-572 1000 mL Evaporating Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 13-878-570 Supplier Heidolph™ Supplier No. 036301130
Add to Cart
Add to Cart
Five condenser assemblies allow customization of evaporator glassware for any application
  • All glassware can be interchanged among rotary evaporator models
  • Each set includes one 1000 mL evaporating flask and one 1000 mL receiving flask
  • All sets are available with standard or plastic safety-coated glassware (halar coating for G5B and abcite coating for G1B/G3B/G6B)
  • Evaporating flasks and vapor tubes with the standard ground joint NS 24/40 are also available with NS 29/32 ground joints
Specifications
Type G1 Diagonal Condenser with safety coating
For Use With (Equipment) Rotary Evaporator
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov