Learn More
Heidolph™ Large Scale Rotary Evaporator Accessory - Seal
SDP
Catalog No. 13878865 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Vacuum replacement seal for LR20 evaporator
Catalog No. For Use With (Equipment)
13-878-865 Vacuum replacement seal for LR20 evaporator
1 options
Catalog No. 13-878-865 Supplier Heidolph™ Supplier No. 036210040
Add to Cart
Add to Cart
Replacement seal for LR20 Large Scale Rotary Evaporator
  • 55/98 seal
Specifications
Type Evaporator Seal
For Use With (Equipment) Vacuum replacement seal for LR20 evaporator
Taper 55/98
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov