Learn More
Heidolph™ Tension Roller for Polymax 2040, Promax 2020, and Unimax 2010 Shakers
SDP
Catalog No. 13889836 Shop All Heidolph North America Products
Change view
Click to view available options
Diameter (Metric) Inside:
1.4 cm
Catalog No. Diameter (Metric) Inside
13-889-836 1.4 cm
1 options
Catalog No. 13-889-836 Supplier Heidolph™ Supplier No. 036303370
Add to Cart
Add to Cart
For use with Polymax 2040, Promax 2020 and Unimax 2010 Platform Shakers
  • Holds larger vessels in place on shaker
  • Configurations available for use with Promax 2020 or 2040, Rotamax 120, Unimax 2010 and Vibramax 100 shakers
Specifications
Product Type Tension Roller
Material PTFE, Stainless Steel
Diameter (Metric) Inside 1.4 cm
For Use With (Equipment) Polymax 2040, Promax 2020, Unimax 2010 platform shakers
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov