Learn More
Heidolph™ Radleys Carousel 12 Plus Reaction Station
GSA_VASDP
Catalog No. 13900161 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Radleys Carousel 6 Plus, Carousel 12 Plus Reaction Systems
Catalog No. For Use With (Equipment)
13-900-161 Radleys Carousel 6 Plus, Carousel 12 Plus Reaction Systems
1 options
Catalog No. 13-900-161 Supplier Heidolph™ Supplier No. 015862450
Add to Cart
Add to Cart
Request a Quote
Simultaneously heats, stirs, and refluxes up to 12 samples, 1-20 mL in size, under an inert atmosphere
Flexibility is inherent in its design: choose any combination of flask sizes, with different sidearm combinations and accessories. The Carousel itself fits on a standard hotplate stirrer and is compatible with standard rotary and most centrifugal evaporators.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Reaction Station
For Use With (Equipment) Radleys Carousel 6 Plus, Carousel 12 Plus Reaction Systems
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov