Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Radleys Carousel Zinc Plated Wire Rack
SDP

Catalog No. 05403772 Shop All Heidolph North America Products
Change view
Click to view available options
Positions:
36
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
05-403-772 36
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-403-772 Supplier Heidolph™ Supplier No. 015860430
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Carousel zinc plated wire rack for use with Carousel 12 Plus Reaction Station™

Choice of size for 36 x 24 mm, 40 x 24 mm, and 80 x 24 mm tubes.

Specifications

Color Silver
Holds Tubes (24 mm)
Material Zinc-plated Wire
Positions 36
Sterility Non-sterile
For Use With (Equipment) Carousel 12 Plus Reaction Station™
Product Type Wire Rack
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.