Learn More
Heidolph™ Radleys Cooled Carousel 12 Plus, Magnetic Stirring Reaction System
SDP
Catalog No. 13900163 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Standard Magnetic Stirrer
Catalog No. For Use With (Equipment)
13-900-163 Standard Magnetic Stirrer
1 options
Catalog No. 13-900-163 Supplier Heidolph™ Supplier No. 015862460
Add to Cart
Add to Cart
Carousel system accessory that offers cost effective low temperature parallel synthesis from ambient down to -78°C
Innovative Cooled Carousel 12 Plus reservoir is designed to accept the removable reflux/inerting head from the Carousel 12 Plus, allowing for the reaction tubes to be easily and rapidly transferred between heating and cooling bases.
  • Fits directly on a standard magnetic stirrer
  • Robust HDPE cooling reservoir is compatible with a wide range of cooling mixtures, including dry ice/acetone for manually controlled cooling from ambient down to -78°C
  • Features a unique non-drip spout and ergonomically designed handle for easy disposal of waste solvents
  • Insulated foam core maintains low temperatures for long periods while protecting the stirrer from freezing (also reduces condensation and ice formation on outer surfaces)
  • HDPE lid keeps reaction cooler for longer, minimizes condensation and ice formation on tubes (maintaining visibility of the contents) and prevents spitting from the cooling mixture
  • Robust HDPE reservoir is virtually unbreakable
Specifications
For Use With (Equipment) Standard Magnetic Stirrer
Type Reaction Station
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov