Learn More
Heidolph™ Radleys GreenHouse Blowdown Evaporator™ Accessories: Blowdown Pin Plates
SDP
Catalog No. 05403796 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Blowdown Pin Plate, 4 x 2
Long Blowdown Pin Plate, 6 x 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05-403-796 Blowdown Pin Plate, 4 x 2
05-403-794 Long Blowdown Pin Plate, 6 x 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-403-796 Supplier Heidolph™ Supplier No. 015860670
Add to Cart
Add to Cart
For use with GreenHouse Blowdown Evaporator™
Interchangeable pin plates with either 8 or 24 hollow blowdown pins deliver an equal flow of gas to each tube, vial or well
Specifications
For Use With (Equipment) Heidolph Radleys GreenHouse Blowdown Evaporator
Type Blowdown Pin Plate, 4 x 2