Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Radleys GreenHouse Plus Reaction Station Accessories: Reaction Tube
SDP

Catalog No. 05403977 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Reaction Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05-403-977 Reaction Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-403-977 Supplier Heidolph™ Supplier No. 015880850
Add to Cart
Add to Cart

For GreenHouse Plus Reaction Stations

Specifications

For Use With (Equipment) GreenHouse Plus Parallel Synthesizer™
Type Reaction Tube
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.