Learn More
Heidolph™ Radleys GreenHouse Plus Parallel Synthesizer™ Accessories - Septum Mats
SDP
Catalog No. 05403979 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Septum Mat
Septum Mat, PTFE-faced
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05-403-979 Septum Mat
05-403-980 Septum Mat, PTFE-faced
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-403-979 Supplier Heidolph™ Supplier No. 015880870
Add to Cart
Add to Cart
For use with GreenHouse Plus Parallel Synthesizer™
  • Allows additions and withdrawals through sealing mats
  • Silicone Septum Mat: The silicone exhibits excellent resistant to heat and cold as well as offering good resistance to a broad range of solvents and solvent vapours. Can be repeatedly pierced whilst maintaining a gas tight seal.
  • Pre-Slit PTFE Faced Silicone Septum Mat: Manufactured from chromatographically clean silicone elastomer, with a thin PTFE film on both sides and 24 pre-slit positions enabling use of blunt syringe needles.
  • Specifications
    For Use With (Equipment) GreenHouse Plus Parallel Synthesizer
    Type Septum Mat