Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Radleys GreenHouse Plus Reaction Station Accessories: Stirring Bar
SDP

Catalog No. 05403978 Shop All Heidolph North America Products
Change view
Click to view available options
Length (Metric):
15 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
05-403-978 15 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-403-978 Supplier Heidolph™ Supplier No. 015880860
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For GreenHouse Plus Reaction Stations

  • PTFE guard-ring protects against accidental contact with hot base
  • Specifications

    Type Stir Bar
    Shape Cylinder
    Length (Metric) 15 mm
    Diameter (Metric) 24 mm
    Quantity 40/Pk.
    Color White
    Material PTFE
    Autoclavable Autoclavable
    Coating PTFE
    For Use With (Equipment) GreenHouse Plus Parallel Synthesizer™
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.