Learn More
Heidolph™ Radleys Heat-On Multi-Well Block Package with Six Inserts
GREENER_CHOICESDP
Catalog No. 05404155 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Hei Series Magnetic Stirrer
Catalog No. For Use With (Equipment)
05-404-155 Hei Series Magnetic Stirrer
1 options
Catalog No. 05-404-155 Supplier Heidolph™ Supplier No. 015852030
Add to Cart
Add to Cart
Includes
Multi-Well holder, One of each insert (for 25, 50 and 100mL flasks), and adaptor plate for 135mm hotplates
Replace messy oil baths and heating mantles; avoid spills and make chemistry safer, cleaner and faster
Not all block designs are the same; test results show that Heat-On heats up 66% faster and uses 30% less energy than other leading brands of blocks.
  • This unique Multi-well block holder is designed to hold either one or two inserts for flasks or tubes
  • No oil baths or heating mantles required- safer operation with less clean-up
  • Can be used with all common hotplates up to 145 mm in diameter; adapter for 135 mm hotplates included in Heat-On system packages
  • Heats up to 200°C, and even up to 260°C for short periods
  • Solid aluminum for fast and even heat transfer from the hotplate into the sample
  • Lightweight and deep well design for increased surface contact contribute to fast heat transfer with added safety — no sticking or broken flasks to worry about
  • Fluoropolymer coating for superior chemical resistance; optional anodized coating also available
Specifications
Closure Material Aluminum
For Use With (Equipment) Hei Series Magnetic Stirrer
Includes 1 x multi-well holder, 1 x 25 mL, 1 x 50 mL and 12 x 100 mL flask inserts, 1 x adapter plate for 135 mm hotplates
Temperature (Metric) Ambient to 2600°C
Type Heat-On Multi-Well Packages