Learn More
Heidolph™ Radleys Rotary Evaporator Adapters
SDP
Catalog No. p-4835746 Shop All Heidolph North America Products
Change view
Click to view available options
Connection Type:
Rodaviss B29 Socket to B24 Cone
Catalog No. Connection Type
05-403-944 Rodaviss B29 Socket to B24 Cone
1 options
Catalog No. 05-403-944 Supplier Heidolph™ Supplier No. 015880520
Only null left
Add to Cart
Add to Cart
Rotary evaporator adapters for use with Carousel 6 Plus Reaction Station™
Choice of size Rodaviss B29 socket to B24 cone, and 24/40 socket to B24 cone.
Specifications
Connection Type Rodaviss B29 Socket to B24 Cone
Type Rotary Evaporator Adapter
For Use With (Equipment) Carousel 6 Plus Reaction Station™
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov