Learn More
Heidolph™ Radleys™ Universal Clamps and Accessories
SDP
Catalog No. 05404148 Shop All Heidolph North America Products
Change view
Click to view available options
Holds:
Flasks, Condensers, Other Glassware
Catalog No. Holds
05-404-148 Flasks, Condensers, Other Glassware
1 options
Catalog No. 05-404-148 Supplier Heidolph™ Supplier No. 015892160
Add to Cart
Add to Cart
Radleys™ Universal Clamps and Accessories are designed to safely support flasks, condensers and other glassware. They feature adjustable telescopic arms and flexible straps to hold glassware. Designed to be raised or lowered with glassware in-situ.
  • Choice of Velcro or rubber supporting straps
  • Rubber straps grip glassware from 15 to 70 mm in diameter - ideal for lifting vessels
  • 5-way clamp features a slot which allows it to be located on to the support rod from the side
  • Velcro straps are designed specifically not to grip glassware tightly, allowing condensers to slide through the strap, whilst still being safely restrained
  • Optional Viton rubber straps offer superior chemical resistance
Specifications
Holds Flasks, Condensers, Other Glassware
Description Strap securing
For Use With (Equipment) StarFish Work Station
Packaging Quantity 10/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov