Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heidolph™ Tuttnauer™ Lab Line EL-PV-G-WS Autoclave
SDP

Catalog No. p-200104870 Shop All Heidolph North America Products
Change view
Click to view available options
Capacity (Metric):
85 L
160 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01-549-773 160 L
13-890-054 85 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-549-773 Supplier Heidolph™ Supplier No. 023210685
Only null left
Add to Cart
Add to Cart
Request a Quote

Designed for quick heat up times and complete drying, this autoclave includes steam generator, and vacuum pump. No plumbing is required for operation, simply fill the reservoirs and plug it in to start using.

Standard Operating Features:

  • Designed to provide high quality, repeatable performance and accountability for a wide range of applications used in modern laboratories
  • For use with Agar preparation as well as the sterilization of liquids, pipettes, glasses, instruments, and biohazards
  • Equipped with a vacuum pump and a steam generator for all laboratory sterilization needs
  • Compact size allows autoclave to fit on a standard benchtop
  • Immediate Steam (G) and Efficient Air Removal (PV) Steam generator in combination with vacuum (PV G) for complete drying
  • PID (Proportional Integral Differential) pressure control
  • Stores the last 200 cycles in built-in memory
  • Two independent, flexible PT100 temperature sensors prevent boil over of liquids and explosions of bottles
  • Highly efficient drying is achieved by uniformly heating the chamber wall of the benchtop autoclave using a coiled pipe
  • The vacuum pump can be used for post vacuum drying to ensure improved drying of porous loads and hollow instruments such as pipette tips
  • Double locking safety device prevents door from opening while chamber is pressurized
  • Control lock-out switch prevents starting a cycle if door is not properly locked
  • Control system protection prevents door from opening at high pressure and temperature
  • All units include trays and printer
  • Unique design with internal reservoirs allows the unit to operate with or without utility connections to treated water, tap water, and a floor sink
  • Applications: Sterilization of liquids, media, instruments, glassware, bioreactors, clothing, and waste

Specifications

Autoclavable Autoclavable
Amperage 23 Amps
Color White
Capacity (English) 42.27 gal.
Capacity (Metric) 160 L
Depth (Metric) Chamber 74.93 cm
Dimensions (L x W) 46.3 x 34.3 in.
Diameter (English) Chamber 19.7 in.
Depth (English) Chamber 29.5 in.
Diameter (Metric) Chamber 50.04
For Use With (Application) Sterilization
For Use With (Equipment) Instruments, glassware or liquids
Length (English) 34.3 in.
Height (English) Exterior 17 in.
Length (Metric) 87.12 cm
Model 5075 EL-PV-G-WS
Product Type Autoclave
Temperature (English) 221°F to 278°F
Temperature (Metric) 105°C to 137°C
Electrical Requirements 230 V, 3 phase (60 Hz) NEMA L15-30
Height (English) Chamber 43.18 cm
Includes Comes with 3 trays, a tray holder, a base plate, and 2 Buck Boost Transformers
Frequency 60 Hz
Printer Yes - Included
Voltage 230 V
Sterility Non-sterile
Dimensions (L x W x H) 46.3 x 34.3 x 62.1 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.