Learn More
Heidolph™ StarFish Workstation Accessories - Loop Strips
SDP
Catalog No. 05404060 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Heidolph StarFish™ Workstation
Catalog No. For Use With (Equipment)
05-404-060 Heidolph StarFish™ Workstation
1 options
Catalog No. 05-404-060 Supplier Heidolph™ Supplier No. 015890330
Add to Cart
Add to Cart
Accessories for use with Heidolph StarFish Workstation
  • These spare velcro straps are designed for use with the StarFish five-way clamp system.
  • They facilitate the secure fastening of assemblies, such as reflux condensers and Soxhlet apparatus, to the StarFish support rod while allowing for unrestricted vertical adjustment.
  • Material: Velcro
  • Quantity: 5 pieces
  • Compatibility: StarFish support system
Specifications
For Use With (Equipment) Heidolph StarFish™ Workstation
Type Hook-and-Loop Strip