Learn More
Heidolph™ StarFish™ Work Station Accessories - Manifolds
GSA_VASDP
Catalog No. 05404062 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Gas/Vacuum Distribution Manifold
Water Distribution Manifold
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05-404-062 Gas/Vacuum Distribution Manifold
05-404-061 Water Distribution Manifold
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-404-062 Supplier Heidolph™ Supplier No. 015890350
Add to Cart
Add to Cart
Accessories for use with Heidolph StarFish™ Work Station
  • Gas/Vacuum Manifold - Even distribution to up to five positions or vessels
  • Water Manifold - distributes water to up to five condensers simultaneously
  • Quick-release connectors
  • Leak-proof shut-off valves
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Specifications
    For Use With (Equipment) Heidolph StarFish Work Station
    Type Gas/Vacuum Distribution Manifold