Learn More
Heidolph™ StarFish™ Work Station Accessories - Support Straps
SDP
Catalog No. 05404150 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Silicone Support Strap
Viton Support Strap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
05-404-150 Viton Support Strap
05-404-149 Silicone Support Strap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-404-150 Supplier Heidolph™ Supplier No. 015892180
Add to Cart
Add to Cart
Accessories for use with Heidolph StarFish™ Work Station
  • Rubber straps grip glassware from 15 to 70 mm in diameter - ideal for lifting vessels
  • Optional Viton rubber straps offer superior chemical resistance.
  • Specifications
    For Use With (Equipment) Heidolph StarFish Work Station
    Length (English) 7.87 in.
    Length (Metric) 200 mm
    Type Viton Support Strap
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov