Learn More
Heidolph™ StarFish Workstation Accessories - Self Adhesive Velcro Pads
Shop All Heidolph North America Products

Description
- Replacement adhesive pads for Starfish telescopic arms.
- Material: Velcro
- Color: Black
- Pattern: Self-adhesive
- Pieces Included: 10
Specifications
Specifications
| For Use With (Equipment) | Heidolph StarFish™ Workstation |
| Type | Hook-and-Loop Pad |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.