Learn More
Heidolph™ Temperature Probes
SDP
Catalog No. 05405277 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
1000, 2000, and 3000 mL Vessels
250 and 500 mL Vessels
5000 mL Vessels
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
05-405-277 1000, 2000, and 3000 mL Vessels
05-405-276 250 and 500 mL Vessels
05-405-278 5000 mL Vessels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-405-277 Supplier Heidolph™ Supplier No. 015950131
Add to Cart
Add to Cart
Wide choice of PTFE Pt100 temperature probes to suit your vessel
  • For use with Reactor-Ready&trade Lab Reactor, Reactor-Ready™ Duo Lab Reactor, and Reactor-Ready™ Pilot Lab Reactor
Specifications
Type Temperature Probe
For Use With (Equipment) 1000, 2000, and 3000 mL Vessels
Connector Types Lemo