Learn More
Heidolph™ Tuttnauer™ Autoclave Accessory, Chamber Brite Autoclave Cleaner
SDP
Catalog No. 13878520 Shop All Heidolph North America Products
Change view
Click to view available options
For Use With (Equipment):
Tuttnauer benchtop sterilizer
Catalog No. For Use With (Equipment)
13-878-520 Tuttnauer benchtop sterilizer
1 options
Catalog No. 13-878-520 Supplier Heidolph™ Supplier No. 023212803
Add to Cart
Add to Cart
Cleaner keeps sterilizer clean and free of water deposit buildup.
  • Fast, convenient and effective way to clean and descale steam sterilizers, including chamber, reservoir and tubing
  • Convenient, premeasured packets replace messy liquids, measuring and scrubbing
  • Simply sprinkle packet into chamber and run
  • Regular use optimizes autoclave function and extends operating life
Specifications
For Use With (Application) Clean and descale steam sterilizers
For Use With (Equipment) Tuttnauer benchtop sterilizer
Product Type Sterilizer Cleaner
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov