Learn More
Seca Corp 206 Mechanical wall-mounted measuring tape, IN
SDP
Supplier:  Seca Corp 2061817139
Encompass_Preferred
The space-saving solution with roll-up mechanism. Uncomplicated fixing on wall with just one screw. With the display window in the head piece, it’s easy to read the height. The durable tape is made of metal. • Space-saving, roll-up mechanism • Simple read-out of results through display window • Durable tape made of metal • Measuring range: 0 – 87 in • Graduation: 1/8 in
Catalog No. NC0429590