Learn More
Sklar Instruments Heinlein Expressor, Sklar 5-1/2 in., Straight, Sharp / Blunt, Flat Serrated Handle, Stainless Steel, EA.
SDP
Catalog No. 501181261 Shop All Sklar Instruments Products
Encompass_Preferred
Heinlein Expressor, Sklar. 5-1/2 in., Straight, Sharp / Blunt, Flat Serrated Handle, Stainless Steel. Sklar's Heinlein Expressor is a dermal instrument that features a lancet end for opening closed comedones and one oval spoon shaped end with a hole for expressing comedones.
Catalog No. 50-118-1261 Supplier Sklar Instruments Supplier No. 064186
Heinlein Expressor, Sklar. 5-1/2 in., Straight, Sharp / Blunt, Flat Serrated Handle, Stainless Steel. Sklar's Heinlein Expressor is a dermal instrument that features a lancet end for opening closed comedones and one oval spoon shaped end with a hole for expressing comedones.