Learn More
Eurofins Discoverx PathHunter HEK 293 ADRA1D β-Arrestin Cell line, 2 vials. (93-0983C1)
Catalog No. 502001745
Encompass
The PathHunter ADRA1D cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1D GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA1D and EA-tagged β-Arrestin-2. Activation of ADRA1D stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1745 Supplier Eurofins Discoverx Supplier No. 930983C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter ADRA1D cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. epinephrine and norepinephrine) based activation of ADRA1D GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged ADRA1D and EA-tagged β-Arrestin-2. Activation of ADRA1D stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.