Learn More
Eurofins Discoverx PathHunter HEK 293 BDKRB2 β-Arrestin Cell Line, 2 vials. (93-0189C1)
Catalog No. 501999890
Encompass
The PathHunter BDKRB2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bradykinin) based activation of BDKRB2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged BDKRB2 and EA-tagged β-Arrestin-2. Activation of BDKRB2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9890 Supplier Eurofins Discoverx Supplier No. 930189C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter BDKRB2 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. bradykinin) based activation of BDKRB2 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged BDKRB2 and EA-tagged β-Arrestin-2. Activation of BDKRB2 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.