Learn More
Eurofins Discoverx PathHunter HEK 293 CCR7 β-Arrestin Cell Line, 2 vials. (93-0195C1)
Catalog No. 501999921
Encompass
The PathHunter CCR7 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL19/MIP-3β) based activation of CCR7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR7 and EA-tagged β-Arrestin-2. Activation of CCR7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9921 Supplier Eurofins Discoverx Supplier No. 930195C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter CCR7 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CCL19/MIP-3β) based activation of CCR7 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged CCR7 and EA-tagged β-Arrestin-2. Activation of CCR7 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.