Learn More
Eurofins Discoverx PathHunter HEK 293 cyno CXCR1 β-Arrestin Cell Line, 2 vials. (93-0980C1)
Catalog No. 502001733
Encompass
The PathHunter cyno CXCR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL8/IL8) based activation of CXCR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno CXCR1 and EA-tagged β-Arrestin-2. Activation of CXCR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cynomolgus monkey version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1733 Supplier Eurofins Discoverx Supplier No. 930980C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter cyno CXCR1 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. CXCL8/IL8) based activation of CXCR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged cyno CXCR1 and EA-tagged β-Arrestin-2. Activation of CXCR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the Cynomolgus monkey version of this protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.