Learn More
Eurofins Discoverx PathHunter HEK 293 EDG3 β-Arrestin Cell Line, 2 vials. (93-0217C1)
Catalog No. 501999985
Encompass
The PathHunter EDG3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. S1P) based activation of EDG3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG3 and EA-tagged β-Arrestin-2. Activation of EDG3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the cynomolgus version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-199-9985 Supplier Eurofins Discoverx Supplier No. 930217C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter EDG3 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. S1P) based activation of EDG3 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged EDG3 and EA-tagged β-Arrestin-2. Activation of EDG3 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. Please note that this product expresses the cynomolgus version of the protein. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.