Learn More
Eurofins Discoverx PathHunter HEK 293 FFAR1 β-Arrestin-1 Cell Line, 2 vials. (93-0977C1)
Catalog No. 502001727
Encompass
The PathHunter FFAR1 (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of FFAR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged FFAR1 and EA-tagged β-Arrestin-2. Activation of FFAR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1727 Supplier Eurofins Discoverx Supplier No. 930977C1
Add to Cart
Add to Cart
Request a Quote
The PathHunter FFAR1 (ARR1) cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. free fatty acids) based activation of FFAR1 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged FFAR1 and EA-tagged β-Arrestin-2. Activation of FFAR1 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.