Learn More
Eurofins Discoverx PathHunter HEK 293 GPR45 β-Arrestin Orphan GPCR Cell Line, 2 vials. (93-0374C1A)
Supplier:  Eurofins Discoverx 930374C1A
Encompass
The PathHunter GPR45 cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand based activation of GPR45 GPCR activity via detection of β-arrestin recruitment. The included cell line overexpresses ProLink-tagged GPR45 and EA-tagged β-Arrestin-2. Activation of GPR45 stimulates the recruitment of β-Arrestin-2 and produces EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. This is an orphan receptor without a known ligand. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-0388
Add to Cart
Request a Quote