Learn More
Cygnus Technologies HEK 293 HCP ELISA KIT, 3G
Supplier:  Cygnus Technologies F650S
Encompass
HEK 293 HCP ELISA Kit, 3G; This kit replaces previous kit F650R; Storage:Standards -10°C to -30°C upon receipt, All other kit reagents 2°C to 8°C; CYGNUS TECHNOLOGIES mfg# F650S
Catalog No. NC1690152