Learn More
Eurofins Discoverx PathHunter HEK 293 KDR/KDR Dimerization Cell Line, 2 vials. (93-0996C1)
Supplier:  Eurofins Discoverx 930996C1
Encompass
The PathHunter KDR/KDR Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. VEGF-121) induced dimerization of KDR/KDR receptors. This product uses EFC technology. The cell line included overexpresses KDR target protein fused to PK alongside KDR target protein fused to EA. Upon ligand-induced dimerization of KDR/KDR, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1770
Add to Cart
Request a Quote