Learn More
ATCC HEK 293 STF; Embryonic Kidney; Human (Homo sapiens)
Catalog No. 502383618
Encompass
HEK 293 STF is an epithelial-like cell line that was isolated in 2002 from the kidney of an embryo. This cell line was stably cotransfected with (1) a 7 x LEF/TCF sites, a minimal thymidine kinase promoter and (2) a pSV2-neo resistance plasmid exhibiting G418 resistance. This cell line was deposited by J Nathans and can be used in assay development research.
Catalog No. 50-238-3618 Supplier ATCC Supplier No. CRL3249
Add to Cart
Add to Cart
HEK 293 STF is an epithelial-like cell line that was isolated in 2002 from the kidney of an embryo. This cell line was stably cotransfected with (1) a 7 x LEF/TCF sites, a minimal thymidine kinase promoter and (2) a pSV2-neo resistance plasmid exhibiting G418 resistance. This cell line was deposited by J Nathans and can be used in assay development research.