Learn More
Eurofins Discoverx PathHunter HEK 293 Tie2 Functional Assay, 2 vials. (93-1024C1)
Supplier:  Eurofins Discoverx 931024C1
Encompass
The PathHunter HEK 293 Tie2 Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated Tie2 dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses ProLink-tagged Tie2 and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/kinases.
Catalog No. 50-200-1872
Add to Cart
Request a Quote