Learn More
Invivogen InvivoGen HEK-Blue™ IL-7 Reporter Cells, Detects Bioactive IL-7, Monitors JAK/STAT5 Pathway, 3 Vials
SDP
Catalog No. NC2155408
Encompass
The InvivoGen HEK-Blue™ IL-7 Reporter Cells are engineered HEK 293 cells designed for detecting and quantifying interleukin-7 (IL-7) through the activation of the JAK/STAT5 signaling pathway. This product offers benefits in screening antibodies and small molecule inhibitors targeting IL-7, ensuring high responsiveness to both human and murine IL-7, setting it apart from standard cell lines.
This item is not returnable. View return policy
Catalog No. NC2155408 Supplier Invivogen Supplier No. HKBIL7
The InvivoGen HEK-Blue™ IL-7 Reporter Cells are engineered HEK 293 cells designed for detecting and quantifying interleukin-7 (IL-7) through the activation of the JAK/STAT5 signaling pathway. This product offers benefits in screening antibodies and small molecule inhibitors targeting IL-7, ensuring high responsiveness to both human and murine IL-7, setting it apart from standard cell lines.