Learn More
Invivogen HEK-Blue™ Detection is a cell culture medium that allows the detection of SEAP as the reporter protein is secreted by the cells.
SDP
Catalog No. NC9482051
Encompass
HEK-Blue™ Detection contains all the nutrients necessary for cell growth and a specific SEAP color substrate. The hydrolysis of the substrate by SEAP produces a purple/blue color that can be easily detected with the naked eye or measured with a spectrophotometer.
Catalog No. NC9482051 Supplier Invivogen Supplier No. HBDET2
HEK-Blue™ Detection contains all the nutrients necessary for cell growth and a specific SEAP color substrate. The hydrolysis of the substrate by SEAP produces a purple/blue color that can be easily detected with the naked eye or measured with a spectrophotometer.