Learn More
Invivogen HEK-Blue™ hTLR7 Cells, NF-κB–SEAP Reporter Line, TLR7-Dependent Signaling, 3-7 x 10^6 cells
SDP
Catalog No. NC2683243
Encompass
HEK-Blue™ hTLR7 cells are engineered HEK293 cells designed for TLR7 receptor signaling studies. They contain a robust NF-κB–SEAP reporter system, enabling precise monitoring of NF-κB activation upon TLR7 stimulation. Ideal for researchers, they facilitate the investigation of TLR7 agonists and inhibitors, offering distinct advantages over standard cell lines due to their unique modifications, including a stable response to specific ligands.
Catalog No. NC2683243 Supplier Invivogen Supplier No. HKBHTLR7V2
HEK-Blue™ hTLR7 cells are engineered HEK293 cells designed for TLR7 receptor signaling studies. They contain a robust NF-κB–SEAP reporter system, enabling precise monitoring of NF-κB activation upon TLR7 stimulation. Ideal for researchers, they facilitate the investigation of TLR7 agonists and inhibitors, offering distinct advantages over standard cell lines due to their unique modifications, including a stable response to specific ligands.