Learn More
Invivogen HEK-Blue™ IL-19/IL-20 Cells, 3-7 x 10e6 cells
SDP
Catalog No. NC3315298
Encompass
Human and murine IL-19, IL-20 & IL-24 cytokine HEK293 reporter cells
Catalog No. NC3315298 Supplier Invivogen Supplier No. HKBIL1920
Human and murine IL-19, IL-20 & IL-24 cytokine HEK293 reporter cells