Learn More
Invivogen HEK-Blue Null1 Cells, 3-7 x 10e6 cells
SDP
Supplier:  Invivogen HKBNULL1
Encompass
HEK-Blue Null1 cells express the SEAP reporter gene under the control of the IFN-beta minimal promoter fused to five NF-KB and AP-1 binding sites.
Catalog No. NC0122468